Learn More
Aviva Systems Biology Corp METTL1 Antibody - middle region (ARP48665_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48665_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against METTL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human METTL1. Alternate names: C12orf1, TRM8, YDL201wApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53560