Learn More
Sino Biological Recombinant Human LCMT2 Protein (Full Length, GST Tag)
Catalog No. 502574431
Encompass_Preferred
Recombinant full length human LCMT2 protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4431 Supplier Sino Biological Supplier No. L32230G100
Recombinant full length human LCMT2 protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.