Learn More
Aviva Systems Biology Corp METTL2B Antibody - N-terminal region (ARP48836_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48836_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against METTL2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human METTL2B. Alternate names: FLJ11350, FLJ12760, METL, METTL2, METTL2A, PSENIP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59933