Learn More
Aviva Systems Biology Corp METTL3 Antibody - middle region (ARP34590_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34590_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against METTL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human METTL3. Alternate names: M6A, IME4, Spo8, MT-A70ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55467