Learn More
Aviva Systems Biology Corp Mettl4 Antibody - C-terminal region (ARP48892_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48892_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mettl4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human Mettl4. Alternate names: RGD1306451, Mettl4, HsT661, AV296509, 2410198H06Rik, A730091E08RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-386