Learn More
Aviva Systems Biology Corp METTL7A Antibody - N-terminal region (ARP42389_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42389_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against METTL7A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human METTL7A. Alternate names: AAM-B, DKFZP586A0522ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58210