Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Accessories for FiveGo™ pH Meters

Catalog No. p-3640210 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Battery Cover
Carry Bag
Wrist Strap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-915-687 Battery Cover
01-915-689 Wrist Strap
01-915-691 Carry Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-915-687 Supplier Mettler Toledo™ Supplier No. 51302962

For use with FiveGo pH meters.

For use with FiveGo pH meters.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Type Battery Cover
For Use With (Application) pH
Includes O-ring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.