Learn More
Mettler Toledo™ Compact Anti-Static Kit
GSA_VA
Catalog No. 01910930 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Excellence Plus Analytical and Micro Balance
Catalog No. For Use With (Equipment)
01-910-930 Excellence Plus Analytical and Micro Balance
1 options
Catalog No. 01-910-930 Supplier Mettler Toledo™ Supplier No. 30090337
For XP, XSE and XPE analytical balances.
  • Integrable and programmable antistatic device
  • Eliminates electrostatic charges on sample containers
  • Includes ionizer assembly and conductive glass door.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Excellence Plus Analytical and Micro Balance
For Use With (Application) Eliminates electrostatic charges on sample containers
Type Compact Anti-Static Kit