Learn More
Mettler Toledo™ uPlace™ Electrode Holder for SevenCompact™ Meters
Catalog No. 01915113 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
SevenCompact S220 pH/Ion Meter and S230 Conductivity Meter
Catalog No. For Use With (Equipment)
01-915-113 SevenCompact S220 pH/Ion Meter and S230 Conductivity Meter
1 options
Catalog No. 01-915-113 Supplier Mettler Toledo™ Supplier No. 30019823
Add to Cart
Add to Cart
For use with SevenCompact S220 pH/Ion Meter and S230 Conductivity Meter.
  • uPlace electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample
  • This allows for faster measurements and reduces the risk of the sample vessel tipping over or damaging the sensor
TRUSTED_SUSTAINABILITY
Specifications
Type uPlace Electrode Holder
For Use With (Application) uPlace electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample
For Use With (Equipment) SevenCompact S220 pH/Ion Meter and S230 Conductivity Meter