Learn More
Mettler Toledo™ Balance Accessories for NewClassic ME: Draft Shields
Catalog No. 01911901 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
NewClassic ME (0.001 g)
NewClassic ME (0.1 mg)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-911-901 NewClassic ME (0.1 mg)
01-911-900 NewClassic ME (0.001 g)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-911-901 Supplier Mettler Toledo™ Supplier No. 30037731
Add to Cart
Add to Cart
Draft Shields for NewClassic ME.
  • Available in low and high varieties
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) NewClassic ME (0.1 mg)
Includes Mounted front and rear Glass panels, sliding doors