Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ 413/IP67 pH Electrode

Catalog No. 01911251 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
BNC, RCA IP67
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01-911-251 BNC, RCA IP67
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-911-251 Supplier METTLER TOLEDO™ Supplier No. 52000379
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A durable, plastic body pH electrode ideal for a wide range of applications that is able to withstand wet and demanding environments (IP67 rated).

  • For use with METTLER TOLEDO portable meters, SevenExcellence and SevenCompact as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Emulsions
  • Suspensions
  • Wastewater
  • Effluent water
  • Proteins & Sulfides
  • General lab

Accessories & Care

  • Open junction and Non-refillable - no additional electrolyte required
  • Non-IP67 rated electrode available: InLab Expert Pro (51343101)
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC, RCA IP67
Junction Open
Material PEEK
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 60°C
Type Combination Electrode; Integrated Temperature Probe
Filling Solution Type Polymer
For Use With (Application) Ideal for emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides
For Use With (Equipment) Fixed Cable to sensor
Internal Reference ARGENTHAL
Model InLab™ 413
Temperature (English) Operating 0°F to 140°F
Length (English) 4.7 in.
Length (English) Cable 39 in.
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.