Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Buffer Solutions: Standard pH

Catalog No. 01913212 Shop All METTLER TOLEDO Products
Change view
Click to view available options
pH Range:
4
4.006
7
10.012
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
01-913-212 4.006
01-913-215 10.012
01-913-209 7
01-915-646 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-913-212 Supplier Mettler Toledo™ Supplier No. 51350052
Add to Cart
Edge
Add to Cart

Storage and refill solutions for pH electrodes

  • Completely miscible
  • Comes with Quality inspection certificate
  • NIST traceable
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point ∼100°C
Color Clear
Connector Types DIN
Type Buffer Solution
Filling Solution Type pH Buffer
Includes Quality inspection certificate
Packaging 250 mL Bottle
pH Range 4.006
Water Soluble Completely Miscible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.