Learn More
Description
- Completely miscible
- Comes with Quality inspection certificate
- NIST traceable
Specifications
Specifications
| Boiling Point | ∼100°C |
| Color | Clear |
| pH | 10.012 |
| Solution Type | pH Buffer |
| Packaging | 250 mL Bottle |
| Solubility Information | Completely Miscible in Water |
| Includes | Quality inspection certificate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.