Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ by Labconco™ Balance Enclosures: Holders

Catalog No. 01913720 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Product Type:
Bottle Holder
Tissue Holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
01-913-720 Tissue Holder
01-913-719 Bottle Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-913-720 Supplier Mettler Toledo™ Supplier No. 63059016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Increase safety and efficiency

These accessories help ensure cleanliness in the lab and reduce the risk of exposure.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Balance Enclosure
Product Type Tissue Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.