Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ by Labconco™ Balance Enclosures: Holders

Catalog No. p-4898557 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Product Type:
Bottle Holder
Tissue Holder
Catalog No. Product Type
01-913-719 Bottle Holder
01-913-720 Tissue Holder
2 options

Catalog No. 01-913-719

Supplier: Mettler Toledo™ 63059015

Only null left
Add to Cart
Add to Cart

Increase safety and efficiency

These accessories help ensure cleanliness in the lab and reduce the risk of exposure.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Balance Enclosure
Product Type Bottle Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.