Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler-Toledo™ Calibration Substance ME 18555, For the calibration of the thermosystem 900, MilliporeSigma™ Supelco™

Catalog No. 111019364 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9364 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9364 Supplier MilliporeSigma Supelco Supplier No. 739835GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzoic acid is an aromatic acid, which occurs physically as colorless leaflets or white needles.

Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 1061°F
CAS 65-85-0
Vapor Pressure 10 mmHg (132°C)
Vapor Density 4.21 (vs. air)
Flash Point 121°C
Physical Form Neat
Linear Formula C6H5 COOH
MDL Number MFCD00002398
Melting Point 121°C to 125°C
Boiling Point 249°C
Synonym Benzoic acid
Type Analytical Standard
Quantity 5 g
Formula Weight 122.12
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.