Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler-Toledo Calibration substance ME 30034252, Phenyl salicylate traceable to primary standards, MilliporeSigma™ Supelco™

Catalog No. 111012695 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2695 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2695 Supplier MilliporeSigma Supelco Supplier No. 447705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 118-55-8
Flash Point 113°C
Physical Form Neat
Linear Formula 2-(HO)C6H4 CO2C6H5
MDL Number MFCD00002213
Melting Point 41°C to 43°C
Boiling Point 172°C to 173°C (12 mmHg)
Synonym Phenyl salicylate; Salol
Type Analytical Standard
Quantity 5 g
Formula Weight 214.22
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.