Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler-Toledo™ Calibration substance ME 51143095, Potassium nitrate traceable to primary standards (LGC), MilliporeSigma™ Supelco™

Catalog No. 111019315 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9315 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9315 Supplier MilliporeSigma Supelco Supplier No. 491435G
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line Mettler-Toledo
CAS 7757-79-1
Physical Form Neat
Linear Formula KNO3
MDL Number MFCD00011409
Melting Point 334°C
Synonym Potassium nitrate
Type Analytical Standard
Quantity 5 g
Formula Weight 101.10
Grade Analytical Standard
Show More Show Less

Mettler-Toledo is a registered trademark of Mettler-Toledo, Inc.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.