Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Interchangeable Buret, for Mettler Toledo™ Titrators

Catalog No. 01911100 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Volume (Metric):
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-911-100 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-911-100 Supplier Mettler Toledo™ Supplier No. 51107500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with V20 Karl Fischer and G20 Compact Titrator.

  • Volume: 5mL
TRUSTED_SUSTAINABILITY

Specifications

Type Titration Burette
Volume (Metric) 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.