Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Replacement Tubing for DL31 Karl Fischer™ Titrators

Catalog No. 01911132 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Solvent Delivery/Drain Tube with Screw Fitting
Tubing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-911-132 Tubing
01-911-128 Solvent Delivery/Drain Tube with Screw Fitting
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-911-132 Supplier Mettler Toledo™ Supplier No. 51107490
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement parts for use with DL31 Karl Fischer titrators.

For use with DL31 Karl Fischer Titrators.
TRUSTED_SUSTAINABILITY

Specifications

Type Tubing
For Use With (Application) Titration
Length (English) 17.7 in.
Length (Metric) 450 mm
Material Silicone
For Use With (Equipment) Mettler Toledo DL31 KF titrator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.