Learn More
Mettler Toledo™ DO308 Karl Fischer™ Drying Oven: Aluminum Inserts
Catalog No. 01912515 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
DO308 Karl Fischer Drying Oven Sample Boats
Catalog No. For Use With (Equipment)
01-912-515 DO308 Karl Fischer Drying Oven Sample Boats
1 options
Catalog No. 01-912-515 Supplier Mettler Toledo™ Supplier No. 51108649
Add to Cart
Add to Cart
For DO308 Karl Fischer drying oven sample boats.
  • Set of 50
TRUSTED_SUSTAINABILITY
Specifications
Type Insert
Material Aluminum
For Use With (Equipment) DO308 Karl Fischer Drying Oven Sample Boats