Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Battery Cover for EL20/EL30 Conductivity Meters

Catalog No. 01915688 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EL-20/EL-30 meter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-915-688 EL-20/EL-30 meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-915-688 Supplier Mettler Toledo™ Supplier No. 51302963

Accessory for use with EL20/EL30 Conductivity Meters.

Accessory for use with EL20/EL30 Conductivity Meters.
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Type Battery Cover
For Use With (Application) pH
Includes O-ring
For Use With (Equipment) EL-20/EL-30 meter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.