Learn More
Mettler Toledo™ Electrode Cable for Karl Fischer™ Titrator
Catalog No. 01912341 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
Lemo
Catalog No. Connector Types
01-912-341 Lemo
1 options
Catalog No. 01-912-341 Supplier Mettler Toledo™ Supplier No. 89602
Add to Cart
Add to Cart
Accessory for use with V10/20S and G10/20S Compact Titrators.
  • Type: SC-Lemo
  • For use with standard and Plug and Play electrodes
  • For V10/20S (01-914-315/ 01-914-315) or G10/20S (01-914-318/ 01-914-319) series titrators
  • Length: 100cm
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Lemo
For Use With (Equipment) Standard and Plug and Play Electrodes
For Use With (Application) Connection to the titrators
Includes S12 grey connection
Length (Metric) Cable 100 cm