Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Electrode Fill Solutions

Catalog No. 01911514 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Filling Solution Type:
1M KCl; 25 mL
FRISCOLYT-B
KCl saturated with AgCl; 25mL
For Use With (Equipment):
DX Series ISE Half Cell
Reference Electrode
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filling Solution Type For Use With (Equipment)
01-911-514 1M KCl; 25 mL DX Series ISE Half Cell
01-911-511 FRISCOLYT-B Reference Electrode
01-911-513 KCl saturated with AgCl; 25mL Reference Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-911-514 Supplier Mettler Toledo™ Supplier No. 51343181
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Made from the highest quality reagents to provide optimum performance.

  • Aqueous solution containing inorganic and organic components
  • Supplied in 25mL (0.9 oz.) bottle

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 0.9 oz.
Capacity (Metric) 25 mL
Concentration 1 mol/L
For Use With (Equipment) DX Series ISE Half Cell
Filling Solution Type 1M KCl; 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.