Learn More
Mettler Toledo™ ErgoClips™ Smart Prep Micro Container Holders
Catalog No. 01913900 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Smart Prep Tare Container
Catalog No. For Use With (Equipment)
01-913-900 Smart Prep Tare Container
1 options
Catalog No. 01-913-900 Supplier Mettler Toledo™ Supplier No. 30138674
Add to Cart
Add to Cart
Save time, increase safety and eliminate transfer errors.
  • Enables one-step dosing directly into Mettler Toledo Smart Prep tare container
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Smart Prep Tare Container
For Use With (Application) Enables one-step dosing directly into Mettler Tole
Type Tare Container Holder