Learn More
Mettler Toledo™ ErgoClips™ Small Basket For Weighing
Catalog No. 01914085 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Test Tubes, Centrifuge Tubes or Small Plastic Tube
Catalog No. For Use With (Equipment)
01-914-085 Test Tubes, Centrifuge Tubes or Small Plastic Tube
1 options
Catalog No. 01-914-085 Supplier Mettler Toledo™ Supplier No. 11106747
Add to Cart
Edge
Add to Cart
For use with test tubes, centrifuge tubes or small plastic tubes of XP, XS, XA analytical and XP, XS micro balance.
The ErgoClips safer dosing method substantially reduces contamination risks, enables fast dosing processes and increases productivity.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Test Tubes, Centrifuge Tubes or Small Plastic Tube
Length (English) 1.2 in.
Length (Metric) 3.2 cm
Width (English) 1.2 in.
Width (Metric) 3.2 cm
Type ErgoClip™ Small Basket