Learn More
Mettler Toledo™ ErgoClips™ Small Flask
Catalog No. 09320362 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For small flask
Catalog No.
09-320-362 For small flask
1 options
Catalog No. 09-320-362 Supplier Mettler Toledo™ Supplier No. 11140180
Add to Cart
Add to Cart
Includes
Disposable Aluminum pans, 50 weighing boats, weigh boat attachment, round-bottom flask holder and test tube holder
For use with XP, XS, XA analytical and XP, XS micro balance.
The ErgoClips safer dosing method substantially reduces contamination risks, enables fast dosing processes and increases productivity.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 1 to 10 mL Volumetric Flask of XP, XS, XA Analytical Balance
Type ErgoClip™ Small Flask