Learn More
Mettler Toledo™ ErgoClips™ Solution Kits
Catalog No. p-4377416 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Analytical Balance
XP26 and XP56 Micro Balance
Includes:
Min weigh micro door, tube, micro flask, including
Minweigh door, flask, tube, small flask, round bot
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Includes
09-320-360 XP26 and XP56 Micro Balance Min weigh micro door, tube, micro flask, including
09-320-361 Analytical Balance Minweigh door, flask, tube, small flask, round bot
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-320-360 Supplier Mettler Toledo™ Supplier No. 11140253
Allow easy weighing directly into tare containers. Mettler Toledo ErgoClips Solution Kit includes all necessary tools for direct dosing with storage case.
  • Guarantees secure placement of different tare containers
  • Dosing is fast and safe thanks to the adjustable MinWeight Door
  • Accurate weighing for reliable, complete sample transfer
Solution Kit Contents:
  • ErgoClip Flask
  • ErgoClip Tube
  • ErgoClip Flask (Small)
  • ErgoClip Round Bottom Flask
  • ErgoClip Basket (Small)
  • ErgoClip Weighing Boat
  • ErgoClip Titration Basket
  • ErgoClip Quantos and vial adapters
  • ErgoClip Filter holder
  • MinWeight Door
  • Stand
  • Various tare containers
  • Spatula
  • Tweezers
Solution Kit Micro Contents:
  • MinWeight Door micro
  • ErgoClip Tube (without holder)
  • ErgoClip Flask micro, including extension
  • ErgoClip Basket micro
  • Stand for ErgoClips micro
  • SmartGrid
  • Tweezers
  • 5mL volumetric flasks
  • Reagent tube, small
  • 1.5 and 0.5mL centrifuge tube
  • 2mL HPLC tube
  • Micro spatula
Solution kit for use with analytical balances.
Micro kit for use with micro balances XP26 or XP56.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XP26 and XP56 Micro Balance
Includes Min weigh micro door, tube, micro flask, including
Type ErgoClip™ Solution Kit