Learn More
Mettler Toledo™ ErgoClips™ Tube for Weighing
Catalog No. 01929112 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
PCR Tubes, Eppendorf Tubes, Reaction Tubes of XP
Catalog No. For Use With (Equipment)
01-929-112 PCR Tubes, Eppendorf Tubes, Reaction Tubes of XP
1 options
Catalog No. 01-929-112 Supplier Mettler Toledo™ Supplier No. 11106784
Add to Cart
Add to Cart
For use with XP, XS, XA analytical and XP, XS micro balance.
The ErgoClips safer dosing method substantially reduces contamination risks, enables fast dosing processes and increases productivity.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) PCR Tubes, Eppendorf Tubes, Reaction Tubes of XP
Type ErgoClip™ Tube