Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Reference Electrodes

Catalog No. p-3640211 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Junction:
Ceramic
PTFE Sleeve
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Junction
01-911-495 PTFE Sleeve
01-911-494 Ceramic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-911-495 Supplier METTLER TOLEDO™ Supplier No. 51343191
Only null left
Add to Cart
Add to Cart

A reference electrode to be paired with half-cell measurement electrodes.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • For use with half-cell measuring electrodes

Accessories & Care

  • S7 connector type: Cable S7 to 2 mm Banana (30281923) required
  • InLab Reference: ceramic junction
  • InLab Reference Plus: moveable PTFE sleeve junction
  • InLab Reference Flow: triple ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types S7
Junction PTFE Sleeve
Material Glass
Temperature (Metric) Operating 0°C to 60°C
Type Half-Cell Reference Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) For routine applications and measurements in Ion-selective half cells
Internal Reference AAg/AgCl
Model InLab™ Reference Plus
Reference Electrolyte Gel
Temperature (English) Operating 32°F to 140°F
Temperature (Metric) Range 0°C to 60°C
Length (English) 4.72 in.
Length (Metric) 12 cm
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 1.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.