Learn More
METTLER TOLEDO™ InLab™ Versatile pH Electrode Replaceable Ceramic Junctions
Catalog No. 01913204 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Junction:
Ceramic
Catalog No. Junction
01-913-204 Ceramic
1 options
Catalog No. 01-913-204 Supplier METTLER TOLEDO™ Supplier No. 51340268
Add to Cart
Add to Cart
Set of 10 ceramic junctions.
  • Accessory
  • For use with InLab™ Versatile and InLab™ Versatile Pro
TRUSTED_SUSTAINABILITY
Specifications
Junction Ceramic
Temperature (Metric) Operating 0°C to 60°C
Type Set of replaceable junctions
For Use With (Application) Alternative for measurements in simple samples, offer an outstanding price/performance ratio
For Use With (Equipment) InLab designations versatile and versatile Pro pH electrode
Includes 10 Ceramic junction