Learn More
METTLER TOLEDO pH Sensor InPro 3253 225/PT1000
Catalog No. NC1788528 Shop All METTLER TOLEDO Products
Encompass
The InPro 3253 225/PT1000 is a pre-pressurized, liquid filled, low maintenance pH sensor, combined with a PT1000 temperature sensor, for in-line measurement in demanding applications. The 225 mm version is optimal for medium installation lengths.
Catalog No. NC1788528 Supplier METTLER TOLEDO Supplier No. 52002569
The InPro 3253 225/PT1000 is a pre-pressurized, liquid filled, low maintenance pH sensor, combined with a PT1000 temperature sensor, for in-line measurement in demanding applications. The 225 mm version is optimal for medium installation lengths.