Learn More
Mettler Toledo™ MA130 Portable pH/Ion Meter
Catalog No. 0192012 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Includes:
Field Kit
IP67-rated Electrode
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-920-12 Field Kit
01-920-10 IP67-rated Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-920-12 Supplier Mettler Toledo™ Supplier No. MA130FK
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Rugged and durable for use in both lab and field environments—so reliable, a battery change every 500 hours is the only internal maintenance it'll ever need. Ultrasonically welded casings are fully waterproofed to IP67 specifications—when used with special booted electrodes, the meter is dustproof and watertight to a depth of 1m for 30 minutes.
Measures pH (range -2.000 to +19.999) and ion concentration (range 0.000 to 19,999) with three-decimal-place precision. Can be used with most ISEs on the market. Also offers NTC and PT1000 temperature measurement, 0- to 9-hour last calibration reminder, adjustable slope and offset limits, and auto-increment sample ID.
Order Info
Packaging Options. Three packages are available; all packages include pH meter, batteries, instructions and an instrument sealing kit. The B (basic) package offers the meter without an electrode. The BE package includes an InLab™ 413 combination pH electrode with integrated temperature sensor and pH buffer sachets. Field Kits (FK) include the InLab combination electrode, pH buffer sachets, and a carrying case.
TRUSTED_SUSTAINABILITY
Specifications
Range (pH) -2 to +19.999
Includes Field Kit
Product Type Portable pH/Ion Meter