Learn More
Mettler Toledo™ MC126 Portable Conductivity Meter
Catalog No. 0192020 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Length (Metric) Cable:
10 m
2 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Cable
01-920-20 10 m
01-920-13 2 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-920-20 Supplier Mettler Toledo™ Supplier No. MC12610M
Includes
meter with mini-DIN, IP67-rated conductivity probe with 6 1/2 ft. (2m) or 32 ft. (10m) cable, sturdy plastic carrying case, batteries, instructions, instrument sealing kit, and premeasured packets of conductivity standard.
For use in lab and field. Offers adjustable parameters—including solids factor and temperature coefficient. Also features serial output and auto-endpoint, auto-off, and display of probe condition. Automatic temperature compensation (ATC) allows for quick probe responses. Memory holds up to 10 calibration readings. Parameters displayed on large LCD.
So reliable, a battery change is the only internal maintenance needed. Ultrasonically welded casing is fully waterproofed to IP67 specifications—when used with supplied probe. Unit is dustproof and watertight to a depth of 3 ft. (1m) for 30 minutes. Weighs just 1 lb. (0.45kg); only 7 3/8L × 1 3/4W × 3 3/8 in.H (20 × 4.5 × 8.5cm), without cable and probe.
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) Cable 10 m