Learn More
Mettler Toledo™ MP Portable pH Meters
Catalog No. 019208 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Includes:
3-in-1 Electrode
Combination pH Electrode
GLP pH Meter Only
IP67 Electrode
Type:
Field Meter kit
GLP Field Meter kit
GLP pH Meter
Portable pH Meter
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Type
01-920-8 Combination pH Electrode GLP Field Meter kit
01-920-2 Combination pH Electrode Portable pH Meter
01-920-3 3-in-1 Electrode Portable pH Meter
01-920-4 Combination pH Electrode Field Meter kit
01-920-5 GLP pH Meter Only GLP pH Meter
01-920-6 IP67 Electrode GLP pH Meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-920-8 Supplier Mettler Toledo™ Supplier No. MP125FK
Includes
All packages include pH meter, batteries, instructions and an instrument sealing kit. The B (basic) package offers the meter without an electrode. The BLE package includes a low-cost, 3-in-1 non-IP67 electrode (01-911-250) and pH buffer sachets; the BE package includes an InLab™ 413 combination pH electrode with integrated temperature sensor and pH buffer sachets. Field Kits (FK) include the InLab combination electrode, pH buffer sachets, and a carrying case.
pH and pH GLP models; Fully waterproofed to IP67 specifications.
Rugged and durable for use in both lab and field environments—so reliable, a battery change every 500 hours is the only internal maintenance they'll ever need. Ultrasonically welded casings are fully waterproofed to IP67 specifications—when used with special booted electrodes, the units are dustproof and watertight to a depth of 1m for 30 minutes.
MP120. Basic pH meter is an excellent value—features pH range of 0.00 to 14.00, electrode condition and slope % indicators, selectable compensated buffers, auto-endpoint and auto-off functions and stable-reading indicator.
MP125. Expands on the capabilities of the MP120 with a pH range of -2.00 to 16.00 and enhanced GLP features including datalogging, date and time stamping, buffer prompting, 0- to 4-hour calibration reminder and last calibration information (including E0 and slope values). Clear visual icons display various functions including stable reading, auto endpoint, date/time, selectable buffers, serial data output, auto-off override and probe condition (immediately after calibration).
TRUSTED_SUSTAINABILITY
Specifications
Includes Combination pH Electrode
Type GLP Field Meter kit