Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ NMR pH Electrode
GSA_VA

Catalog No. 01920167 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
S7
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01-920-167 S7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-920-167 Supplier METTLER TOLEDO™ Supplier No. 59904572
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A pH electrode featuring a slim shaft diameter that allows for measurements in NMR tubes or of very small sample volumes.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • NMR tubes
  • Small volume samples
  • Small measuring vessels

Accessories & Care

  • Ceramic junction
  • Refillable 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Order Info

Metal Half-Cells (01-920-162, -163 and -164) and U-Electrodes (09-320-200 and -201) also available separately.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description pH electrode InLab NMR
Connector Types S7
Junction Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode
Filling Solution Type 3mol/L KCl AgCl saturated
For Use With (Application) For particularly long test tubes, vials or vessels
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference Ag/AgCl
Membrane U Glass
Model InLab™ NMR
Storage Requirements 3mol/L KCl AgCl saturated
Temperature (English) Operating 32°F to 176°F
Temperature (Metric) Range 0°C to 80°C
Length (English) 7.87 in.
Length (Metric) 200 mm
Diameter (English) 0.12 in.
Diameter (Metric) 3 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.