Learn More
Mettler Toledo™ Replacement Parts for V20 Karl Fischer™ Autotitrator
Catalog No. 01910730 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Beaker
Cover
Drying Cup
Drying Tube Seal
Molecular Sieve
O-ring
Screw Cap
Septa
Silicone Tubing
Threaded Ring
Titration Vessel
Visco-Spoon™
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-910-730 Beaker
09-316-106C Screw Cap
09-316-104P Drying Cup
09-316-104R Molecular Sieve
01-911-131 Silicone Tubing
01-911-121 Cover
01-911-130 Drying Tube Seal
01-911-125 O-ring
01-911-134 Titration Vessel
09-313-152B Septa
01-911-133 Threaded Ring
01-910-732 Visco-Spoon™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 01-910-730 Supplier Mettler Toledo™ Supplier No. 51107497
Add to Cart
Add to Cart
For use with V20 Karl Fischer Autotitrators.
  • Can be attached to an external oil or water bath
TRUSTED_SUSTAINABILITY
Specifications
Type Beaker
For Use With (Application) Water content determination
For Use With (Equipment) V20 Karl Fischer titrators