Learn More
Mettler Toledo™ Replacement Parts for V20 Karl Fischer™ Autotitrator
Catalog No. 09316104P Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Beaker
Cover
Drying Cup
Drying Tube Seal
Molecular Sieve
O-ring
Screw Cap
Septa
Silicone Tubing
Threaded Ring
Titration Vessel
Visco-Spoon™
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-316-104P Drying Cup
09-316-106C Screw Cap
09-316-104R Molecular Sieve
01-911-131 Silicone Tubing
01-911-121 Cover
01-911-130 Drying Tube Seal
01-911-125 O-ring
01-911-134 Titration Vessel
09-313-152B Septa
01-911-133 Threaded Ring
01-910-732 Visco-Spoon™
01-910-730 Beaker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 09-316-104P Supplier Mettler Toledo™ Supplier No. 23961
Add to Cart
Edge
Add to Cart
For use with V20 Karl Fischer Autotitrators.
  • Accessory for DV1000 titration burettes
  • For titrant protection from moisture or CO2
  • With cover
TRUSTED_SUSTAINABILITY
Specifications
Type Drying Cup
For Use With (Application) Water content determination
Weight (Metric) 453.5 g