Learn More
Mettler Toledo™ RS-P28 Printers
Catalog No. 01911245 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Electrical Requirements:
100/240 V 50/60 Hz
Catalog No. Electrical Requirements
01-911-245 100/240 V 50/60 Hz
1 options
Catalog No. 01-911-245 Supplier Mettler Toledo™ Supplier No. 11124324
Add to Cart
Add to Cart
Delivers GxP-compliant printouts on regular, adhesive or lightfast paper.
  • Ergonomic and attractive design
  • Fast and reliable with simple/sophisticated applications
  • Supports connected instrument languages
  • Build-in real time clock and calendar combined with lightfast paper for documentation compliant with GxP
  • Statistics and Totaling applications
  • Automatically detects settings of connected device
  • Large paper rolls with fewer refills
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 2.2 in. Wide Paper
Electrical Requirements 100/240 V 50/60 Hz
Includes RS232C, 9 pin male connector
Type Compact Printer for Standard Paper