Learn More
Mettler Toledo™ Expansion Module for SevenExcellence™ DO/BOD Meters
Catalog No. 01910928 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Model:
For SevenExcellence DO/BOD
Catalog No. Model
01-910-928 For SevenExcellence DO/BOD
1 options
Catalog No. 01-910-928 Supplier Mettler Toledo™ Supplier No. 30034474
Add to Cart
Add to Cart
For use with SevenExcellence Dissolved Oxygen and DO/BOD Meters.
The modular design of the SevenExcellence meter line allows you to expand your instrument with additional measurement parameters for future needs at any time. Just add the Dissolved Oxygen (DO) / Biochemical Oxygen Demand (BOD) module, switch on the instrument and the new expansion unit is automatically recognized and ready to use. SevenExcellence fits, in total, three expansion units in any composition.
TRUSTED_SUSTAINABILITY
Specifications
Channels 1
Measurement Modes Dissolved Oxygen
Percent Saturation Range (Polarographic DO) 0 to 600%
Temperature Range (Metric) 0°C to 60°C
Model For SevenExcellence DO/BOD
Product Type Expansion Module