Learn More
Mettler Toledo™ Rubber Holster for SevenGo™ pH Meters
Catalog No. 01917154 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
SevenGo multiparameter meter
Catalog No. For Use With (Equipment)
01-917-154 SevenGo multiparameter meter
1 options
Catalog No. 01-917-154 Supplier Mettler Toledo™ Supplier No. 51302322
Add to Cart
Add to Cart
Accessory for SevenGo pH meter.
  • Elastic
  • Provides impact protection
  • Allows to attach electrode and change batteries without removing protection cover
TRUSTED_SUSTAINABILITY
Specifications
Type Holster
Material Rubber
For Use With (Equipment) SevenGo multiparameter meter