Learn More
Mettler Toledo™ Siphon Tip For Titration Sensor Interchangable Burettes
Catalog No. 01910718 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
DV1000 Titration Burettes
Catalog No. For Use With (Equipment)
01-910-718 DV1000 Titration Burettes
1 options
Catalog No. 01-910-718 Supplier Mettler Toledo™ Supplier No. 23240
Add to Cart
Add to Cart
For dispensing tube for titration sensors interchangable burettes 5mL, 10mL and 20mL.
  • Anti-diffusion
  • Five-piece set
Compliance
ISO 8655-3
TRUSTED_SUSTAINABILITY
Specifications
Type Siphon Tip
For Use With (Equipment) DV1000 Titration Burettes