Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Hydrofluoric pH Electrode

Catalog No. 01911480 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
S7
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01-911-480 S7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-911-480 Supplier METTLER TOLEDO™ Supplier No. 51343176
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A pH electrode featuring a glass body that is resistant to hydrofluoric acid.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Hydrofluoric (HF) bearing sample

Accessories & Care

  • Ceramic junction
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Description pH electrode InLab Hydrofluoric
Connector Types S7
Junction Ceramic
Material Glass
pH Range 1 to 11
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) Improves service life and measurement accuracy at differing sample temperatures
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference ARGENTHAL with Ag+ trap
Membrane HF Glass
Model InLab™ Hydrofluoric
Response Time <20 sec.
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Tip Type Bulb
Length (English) 4.72 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.