Learn More
Mettler Toledo™ Stopper for C20 and C30 Series Karl Fischer™ Titrators
Catalog No. 01910746 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Karl Fischer C20, C30 Series Titrators
Catalog No. For Use With (Equipment)
01-910-746 Karl Fischer C20, C30 Series Titrators
1 options
Catalog No. 01-910-746 Supplier Mettler Toledo™ Supplier No. 51108741
Add to Cart
Edge
Add to Cart
For use with C20 and C30 Series Karl Fischer Titrators.
  • Material: PTFE
  • With septum disk
TRUSTED_SUSTAINABILITY
Specifications
Type PTFE Stopper
Includes Septum disk
Material PTFE
For Use With (Equipment) Karl Fischer C20, C30 Series Titrators