Learn More
Mettler Toledo™ Titration Burette For Titration Sensor
Catalog No. 01911102 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Volume (Metric):
20 mL
Catalog No. Volume (Metric)
01-911-102 20 mL
1 options
Catalog No. 01-911-102 Supplier Mettler Toledo™ Supplier No. 51107502
Add to Cart
Add to Cart
Includes
RFID chip
Features optional ceramic valve discs for crystallizing titrants.
  • Allow rapid titrant change
  • Red protection tubing against light
  • Replaceable PTFE valve discs
  • PTFE pistons w/tripple lip seal assure good tightness
Compliance
ISO 8655-3
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Burette
Includes RFID chip
Volume (Metric) 20 mL