Learn More
Mettler Toledo™ uMix™ Magnetic Stirrer
Catalog No. 01911606 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Model:
Magnetic stirrer uMix
Catalog No. Model
01-911-606 Magnetic stirrer uMix
1 options
Catalog No. 01-911-606 Supplier Mettler Toledo™ Supplier No. 30040000
Add to Cart
Edge
Add to Cart
Provides an extra degree of precision for your applications.
Powered and controlled externally, the magnetic stirrer allows for enhanced reproducibility of results and higher efficiency in the measuring process.
The magnetic stirrer can be combined with either the SevenCompact™ pH/Ion S220, the SevenCompact Conductivity S230, or the multiparameter SevenExcellence™ benchtop meters.
TRUSTED_SUSTAINABILITY
Specifications
Display None
Description Magnetic stirrer uMix
Model Magnetic stirrer uMix