Learn More
Mettler Toledo™ V20 Karl Fischer™ Titrator Interface Cables
Catalog No. 0191892 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connection Type:
DB25 Female
DB9 Female
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
01-918-92 DB25 Female
01-918-93 DB9 Female
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-918-92 Supplier Mettler Toledo™ Supplier No. 11101052
Add to Cart
Add to Cart
Interface cables for use with V20 Karl Fischer Titrator.
  • Connect Mettler Toledo balances to printer
  • For use with: AB, HB, PG-S, PB and SB series balances
  • Length:  39 in.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) AB, HB, PG-S, PB and SB Series Balance
Connection Type DB25 Female
Length (English) 39 in.
Length (Metric) 99.06 cm