Learn More
Mettler Toledo™ V20 Three-Hole Adapter for Titration Stand
Catalog No. 09316101E Shop All METTLER TOLEDO Products
Change view
Click to view available options
Includes:
Standard 10 and 7.5 needle hole and appropriate stopper
Catalog No. Includes
09-316-101E Standard 10 and 7.5 needle hole and appropriate stopper
1 options
Catalog No. 09-316-101E Supplier Mettler Toledo™ Supplier No. 23982
Add to Cart
Edge
Add to Cart
Three-hole adapter for titration stand.
  • For use with V20 and V30 Karl Fischer titrators
  • Includes ST10, ST7.5 and needle hole and pin hole stoppers
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Includes Standard 10 and 7.5 needle hole and appropriate stopper