Learn More
Enzo Life Sciences Metyrapone, (200mg), CAS Number: 54-36-4
SDP
Supplier:  Enzo Life Sciences BMLEI2560200
Encompass_Preferred
Cytochrome P450 inhibitor. Inhibits cytochrome P-450-mediated prostaglandin ω/ω-1 hydroxylase activity. Formula: C14H14N2O. MW: 226.34. CAS Number: 54-36-4. Purity: ≥99%. Appearance: White solid. Solubility: Soluble in water (25 mg/ml), DMSO (25 mg/ml) or ethanol (25mg/ml). Long Term Storage: Ambient
Catalog No. NC9196029