Learn More
Fine Science Tools METZENBAUM BABY SCISSORS
SDP
Catalog No. NC1549194
Encompass
Tips:Blunt-Blunt, Alloy / Material:Stainless Steel,Cutting Edge:35mm,Length:14.5cm,Tip Shape: Straight
This item is not returnable. View return policy
Catalog No. NC1549194 Supplier Fine Science Tools Supplier No. 1401814
Tips:Blunt-Blunt, Alloy / Material:Stainless Steel,Cutting Edge:35mm,Length:14.5cm,Tip Shape: Straight